Anti-Cdh5 Rat Monoclonal Antibody [clone: BV13] (PE (Phycoerythrin))
PRSI76-863
New Product
- Type antilichaam:Primair
- antigeen symbool:Cdh5
- Clonality:Monoklonaal
- Clone:BV13
- Conjugatie:PE (fycoerythrin)
- Flowcytometrie:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Kruisadsorptie:No
- Formulier:liquid
- Gen-ID:P55284
- Antigeen synoniemen:Cdh5
- Opslagbuffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Opslagtemperatuur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- VPE:0,1 mg
The BV13 monoclonal antibody specifically reacts with mouse CD144 (VE-Cadherin), a 120 kDa type II Cadherin. CD144 is a hemophilic adhesion molecule of the endothelial cell line essential to maintaining cell layer integrity. It is involved in cell growth inhibition, migration, and survival.
Type: Primary
Antigen: Cdh5
Clonality: Monoclonal
Clone: BV13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse