Specificaties
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:OKT8
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P01732
- Antigen synonyms:CD8A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:500 Tests
Specificaties
Over dit artikel
The OKT8 monoclonal antibody specifically reacts with human CD8alpha molecule, a type I transmembrane glycoprotein of 32 to 34 kDa. CD8 alpha is a member of the Ig superfamily, expressed as a homodimer (CD8 alpha alpha) or as a heterodimer (CD8 alpha beta). CD8+ alpha beta T lymphocytes express both CD8 alpha alpha and CD8 alpha beta, while some T lymphocytes and the natural killer cells express only the homodimers. CD8 binds to MHC class I and influences the development and the activation of T lymphocytes. OKT8, RPA-T8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitoPEor block each other by steric hindrance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450 nm.
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: OKT8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human