Specifications
- Antibody type:Primary
- Antigen name:lymphocyte antigen 6 complex, locus G
- Antigen symbol:Ly6g
- Clonality:Monoclonal
- Clone:RB6-8C5
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P35461
- Antigen synonyms:Ly6g
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
Specifications
About this item
The RB6-8C5 monoclonal antibody reacts with the mouse Ly-6G (also known as Gr-1). The Ly-6G protein is a myeloid differentiation antigen of 21 to 25 kDa, expressed in a regulated manner by the myeloid lineage in the bone marrow, where the level of antigen expression is correlated with the granulocyte maturation and differentiation. In the bone marrow, the antigen is not expressed by the erythroid cells. From the peripheric cells, RB6-8C5 binds with monocytes, neutrophils and eosinophils.As a marker for the mouse monocytes, macrophages and granulocytes, the RB6-8C5 antibody is usually combined with M1/70, a macrophage labeling antibody (Anti-CD11b), for phenotypic analysis.
Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse