Anti-CD3E Mouse Monoclonal Antibody [clone: UCHT1] (VioletFluor® 450)
Catalogus # PRSI76-284
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD3e molecule, epsilon (CD3-TCR complex)
- Antigen symbol:CD3E
- Clonality:Monoclonal
- Clone:UCHT1
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P07766
- Antigen synonyms:CD3E
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The UCHT1 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3 epsilon. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: VioletFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human