Anti-Il4 Rat Monoclonal Antibody [clone: 11B11] (PE (Phycoerythrin))
Catalogus # PRSI77-062
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 4
- Antigen symbol:Il4
- Clonality:Monoclonal
- Clone:11B11
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P07750
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
Specifications
About this item
The 11B11 monoclonal antibody specifically reacts with mouse 14 kDa cytokine IL-4, expressed by the activated NKT cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen.The 11B11 antibody neutralizes the IL-4 cytokine.
Type: Primary
Antigen: Il4
Clonality: Monoclonal
Clone: 11B11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse