Anti-IFNG Mouse Monoclonal Antibody [clone: NIB42]
Catalogus # PRSI77-053
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:interferon, gamma
- Antigen symbol:IFNG
- Clonality:Monoclonal
- Clone:NIB42
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Neutralisation:Yes
- Format:purified antibody
- Form:liquid
- Gene ID:P01579
- Antigen synonyms:IFNG
- Storage buffer:Phosphate-buffered aqueous solution, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0,01 EU/µg of the protein.
- Pk:0,05 mg
Specifications
About this item
The NIB42 monoclonal antibody specifically reacts with human interferon gamma, a multifunctional cytokine that is produced mainly by activated T and NK cells. Interferon gamma has anti-viral and anti-parasitic properties and plays an important role in the activation, growth, and differentiation of macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-alpha, to inhibit proliferation of normal and transformed cells. The NIB42 antibody neutralizes IFN-gamma.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: NIB42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human