Anti-COL4A1 Rabbit Polyclonal Antibody
PRSI62-860
- Antibody type:Primary
- Antigen name:Collagen alpha-1(IV) chain
- Antigen symbol:COL4A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:27-54 aa
- Form:Liquid
- Gene ID:1282
- Modification:None
- Molecular weight:161 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This COL4A1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 27-54 amino acids from the N-terminal region of human COL4A1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:25.
Type: Primary
Antigen: COL4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 27-54
Host: Rabbit
Isotype: Ig
Reactivity: Human