Anti-ALDH7A1 Rabbit Polyclonal Antibody
PRSI23-693
New Product
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 7 family member A1
- Antigen symbol:ALDH7A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:501
- Antigen synonyms:epididymis secretory sperm binding protein|alpha-aminoadipic semialdehyde dehydrogenase|betaine aldehyde dehydrogenase|EPD|antiquitin-1|PDE|delta1-piperideine-6-carboxylate dehydrogenase|alpha-AASA dehydrogenase|ATQ1|26g turgor protein homolog|P6c dehydrogenase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 430-539 of human ALDH7A1 (NP_001173.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ALDH7A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat