Anti-INSR Recombinant Antibody [Clone: Fab 83-7]
Catalog # PRSI24-498
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:insulin receptor
- Antigen symbol:INSR
- Clonality:Recombinant
- Clone:Fab 83-7
- Conjugation:Unconjugated
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:3643
- Antigen synonyms:CD220
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor
- Tested applications:Crystallography
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: insulin receptor
Clonality: Recombinant
Clone: Fab 83-7
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human