Anti-ITGA4 Recombinant Antibody [Clone: Natalizumab]
Catalog # PRSI24-484
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:integrin subunit alpha 4
- Antigen symbol:ITGA4
- Clonality:Recombinant
- Clone:Natalizumab
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG4 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:3676
- Antigen synonyms:alpha 4 subunit of VLA-4 receptor|CD49D|Integrin alpha-IV|ITGA4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The RAMOS cell line, a non-EBV releasing cell line, injected into mice
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: integrin subunit alpha 4
Clonality: Recombinant
Clone: Natalizumab
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4 kappa
Reactivity: Human