Anti-HSD3B1 Recombinant Antibody [Clone: SAIC-34B-124]
Catalog # PRSI24-332
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
- Antigen symbol:HSD3B1
- Clonality:Recombinant
- Clone:SAIC-34B-124
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:3283
- Antigen synonyms:TDPX1|Peroxiredoxin-2|NKEFB
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide "LSEDYGVLK", derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH
- Tested applications:ImmunoMRM
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Clonality: Recombinant
Clone: SAIC-34B-124
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human