Anti-RAD23B Recombinant Antibody [Clone: SAIC-28A-29]
PRSI24-326
- Antibody type:Primary
- Antigen name:RAD23 homolog B, nucleotide excision repair protein
- Antigen symbol:RAD23B
- Clonality:Recombinant
- Clone:SAIC-28A-29
- Conjugation:Unconjugated
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:5887
- Antigen synonyms:UV excision repair protein RAD23 homolog B
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide "ILNDDTALK", derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH
- Tested applications:ImmunoMRM
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: RAD23 homolog B, nucleotide excision repair protein
Clonality: Recombinant
Clone: SAIC-28A-29
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human