Anti-XPNPEP1 Recombinant Antibody [Clone: SAIC-07B-14]
PRSI24-308
- Antibody type:Primary
- Antigen name:X-prolyl aminopeptidase 1
- Antigen symbol:XPNPEP1
- Clonality:Recombinant
- Clone:SAIC-07B-14
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:7511
- Antigen synonyms:XPNPEPL1|XPNPEPL|Xaa-Pro aminopeptidase 1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide "GSLTFEPLTLVPIQTK" derived from Xaa-Pro aminopeptidase 1 conjugated to KLH
- Tested applications:ImmunoMRM
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: X-prolyl aminopeptidase 1
Clonality: Recombinant
Clone: SAIC-07B-14
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human