Anti-AOC3 Recombinant Antibody [Clone: 2D10]
Catalog # PRSI24-202
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:amine oxidase copper containing 3
- Antigen symbol:AOC3
- Clonality:Recombinant
- Clone:2D10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:8639
- Antigen synonyms:Amine oxidase|copper containing 3|Vascular adhesion protein 1|Membrane primary amine oxidase|Semicarbazide-sensitive|HPAO|copper-containing amine:oxyg|SSAO|AOC3|VAP1|Copper amine oxidase|amine oxidase
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Affinity purified VAP-1 from tonsil stroma of human origin
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: amine oxidase copper containing 3
Clonality: Recombinant
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human