Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD59 Recombinant Antibody [Clone: 193-27]
Anti-CD59 Recombinant Antibody [Clone: 193-27]
Catalog # PRSI24-759
Supplier:  ProSci Inc.
Anti-CD59 Recombinant Antibody [Clone: 193-27]
Catalog # PRSI24-759
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD59 molecule (CD59 blood group)
  • Antigen symbol:
    CD59
  • Clonality:
    Recombinant
  • Clone:
    193-27
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    966
  • Antigen synonyms:
    Protectin|MIRL|MAC-inhibitory protein|HRF-20|1F5|MACIF|HRF20|Membrane inhibitor of reactive lysis|MEM43 antigen|20 kDa homologous restriction factor|CD59 glycoprotein|1F5 antigen|Membrane attack complex inhibition factor|MAC-IP
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizng mice with stimulated human leukocytes
  • Pack type:
    Vial
  • Pk:
    0,05 mg

Specifications

About this item

Type: Primary
Antigen: CD59 molecule (CD59 blood group)
Clonality: Recombinant
Clone: 193-27
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgM kappa
Reactivity: Human