Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-HSD3B1 Recombinant Antibody [Clone: SAIC-34D-11]
  PRSI24-333
Anti-HSD3B1 Recombinant Antibody [Clone: SAIC-34D-11]
  PRSI24-333

 

  • Antibody type:
    Primary
  • Antigen name:
    hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
  • Antigen symbol:
    HSD3B1
  • Clonality:
    Recombinant
  • Clone:
    SAIC-34D-11
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3283
  • Antigen synonyms:
    TDPX1|Peroxiredoxin-2|NKEFB
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Peptide 'DHPFGFVAVPTK', derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KL
  • Tested applications:
    immunoMRM
  • Pack type:
    Vial
  • Pk:
    0,2 mg

 

 

Type: Primary
Antigen: hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Clonality: Recombinant
Clone: SAIC-34D-11
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human