Anti-NFKB2 Recombinant Antibody [Clone: SAIC-26C-15]
Catalog # PRSI24-323
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:nuclear factor kappa B subunit 2
- Antigen symbol:NFKB2
- Clonality:Recombinant
- Clone:SAIC-26C-15
- Conjugation:Unconjugated
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:4791
- Antigen synonyms:Nuclear factor NF-kappa-B p100 subunit|LYT10
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide 'IEVDLVTHSDPPR' derived from the transcription factor NFKB2 (Nuclear factor NF-kappa-B p100 subunit) conjugated to KL
- Tested applications:immunoMRM
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: nuclear factor kappa B subunit 2
Clonality: Recombinant
Clone: SAIC-26C-15
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human