Order Entry
Netherlands
ContactUsLinkComponent
 
Fura 2-AM ≥95% (by HPLC)
 
undefined
Fura 2-AM ≥95% (by HPLC)

 

  • Appearance (Colour):
    Yellow to Yellow-Green
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Yellow to Yellow-Green
  • Solubility (Turbidity)
    10 mg/ml, acetone:
    Clear
  • Purity (HPLC):
    ≥95%

 

Frequently Bought Together