Trypsin
Marketsource
Supplier: Roth Carl
About this item
2 Options Available Below
Product Details & Documents
Documents
ROTH5455.1_TDS_EN
Datasheet
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ROTH5455.1
- ROTH5455.2
- Enzyme name:
- Trypsin
- Trypsin
- MDL No.:
- MFCD00082094
- MFCD00082094
- CAS No.:
- 9002-07-7
- 9002-07-7
- Purity:
- 1× crystallized
- 1× crystallized
- Storage conditions:
- Refrigerator
- Refrigerator
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.ROTH5455.1Enzyme nameTrypsinMDL No.MFCD00082094CAS No.9002-07-7Purity1× crystallizedStorage conditionsRefrigeratorMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ROTH5455.2Enzyme nameTrypsinMDL No.MFCD00082094CAS No.9002-07-7Purity1× crystallizedStorage conditionsRefrigeratorMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



