Order Entry
Netherlands
ContactUsLinkComponent
 
Orthoboric acid 20 g/l in aqueous solution
Catalog # 2733.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthoboric acid 20 g/l in aqueous solution
Catalog # 2733.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    1.9 - 2.1 %

Specification Test Results

About this item

CAS Number:10043-35-3
 :
Formula:B(OH)₃
MDL Number:MFCD00011337
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2