Pesticide reference standard, Diazinon 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA12210000AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:333-41-5
- Description:Diazinon 100 µg/ml in acetonitrile
- Formula:C₁₂H₂₁N₂O₃PS
- Description:Diazinon