Pesticide reference standard, Diazinon 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12210000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:333-41-5
- Description:Diazinon 10 µg/ml in acetonitrile
- Environmentally Preferable:
- Formula:C₁₂H₂₁N₂O₃PS
- Description:Diazinon