Pesticide reference standard, Chlorpyrifos 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL11600000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2921-88-2
- Description:Chlorpyrifos 10 µg/ml in cyclohexane
- Formula:C₉H₁₁Cl₃NO₃PS
- Description:Chlorpyrifos