Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
Corticosterone ≥98% (by HPLC)
  ENZOALX370007M050
 :  
undefined
 
 
Corticosterone ≥98% (by HPLC)
  ENZOALX370007M050
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Purity:
    ≥98% (HPLC)

 

 

 :
 :14,02538
 :346.5 g/mol
 :200-019-6
 :GHS07
 :GHS08
 :Ambient
 :50-22-6
 :C₂₁H₃₀O₄
 :MFCD00037715