Order Entry
Netherlands
ContactUsLinkComponent
Aflatoxin M2 ≥98% (by HPLC, TLC)
Toxic
Health
Irritant
Aflatoxin M2 ≥98% (by HPLC, TLC)
Catalog # ENZOALX630114MC01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Toxic
Health
Irritant
Aflatoxin M2 ≥98% (by HPLC, TLC)
Catalog # ENZOALX630114MC01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Seq:
    1286

Specifications

Specification Test Results

  • Source:
    Isolated from Aspergillus flavus
  • Purity:
    ≥98% (HPLC, TLC)

Specification Test Results

About this item

 :
 :330.3 g/mol
Merck Index:14,00182
Storage Temperature:Freezer
EINECS:229-989-9
UN:3462
ADR:6.1,I
 :GHS06
 :GHS08
 :GHS07
CAS Number:6885-57-0
Formula:C₁₇H₁₄O₇