Order Entry
Netherlands
ContactUsLinkComponent
DAR-4M AM (cell permeable) 5 mM ≥98% (by HPLC)
Irritant
DAR-4M AM (cell permeable) 5 mM ≥98% (by HPLC)
Catalog # ENZOALX620069M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irritant
DAR-4M AM (cell permeable) 5 mM ≥98% (by HPLC)
Catalog # ENZOALX620069M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Concentration:
    ~5 mM
  • Purity:
    ≥98% (HPLC)
  • Formulation:
    Dissolved in 0.32 ml DMSO

Specification Test Results

About this item

 :
 :630.5 g/mol
 :GHS07
Storage Temperature:Freezer
Formula:C₂₈H₃₁N₄O₅·I