

Lipopolysaccharides (from Salmonella minnesota) R595 (Re), TLRgrade™ ready-to-use
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
:
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
- Purity:Absence of detectable protein or DNA contaminants with agonistic TLR activity
Specification Test Results
Frequently Bought Together