Order Entry
Netherlands
ContactUsLinkComponent
(+)-Bicuculline ≥98% (by HPLC)
Toxic
Health
(+)-Bicuculline ≥98% (by HPLC)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Toxic
Health
(+)-Bicuculline ≥98% (by HPLC)
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Appearance:
    Off-white to yellow powder

Specification Test Results

Frequently Bought Together