Order Entry
Netherlands
ContactUsLinkComponent
Aconitine ≥98% (by HPLC)
Toxic
Aconitine ≥98% (by HPLC)
Catalog # ENZOALX550232M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Toxic
Aconitine ≥98% (by HPLC)
Catalog # ENZOALX550232M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Source:
    Isolated from Aconitum karacolium

Specification Test Results

About this item

 :
 :645.746 g/mol
Merck Index:14,00118
Storage Temperature:Refrigerator
EINECS:206-121-7
UN:1544
ADR:6.1,I
 :GHS06
CAS Number:302-27-2
Formula:C₃₄H₄₇NO₁₁
MDL Number:MFCD00082375