Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:115457-83-5
:
Formula:C₁₅H₂₉NO₇
MDL Number:MFCD00077425
:335.397 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity (HPLC):≥98%
- Appearance:White to off-white crytalline solid
Specification Test Results
Frequently Bought Together