Adenosine-5'-diphosphate monopotassium salt ≥87%
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:72696-48-1
Formula:C₁₀H₁₄N₅O₁₀P₂K
MDL Number:MFCD00066472
:465.29 g/mol
Merck Index:14,00154
Storage Temperature:Freezer
Specification Test Results
- Purity (assay):≥87%
- Source:Isolated from yeast
Specification Test Results
Frequently Bought Together