IM-54 ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:861891-50-1
:
Formula:C₁₉H₂₃N₃O₂
MDL Number:MFCD18382109
:325.40 g/mol
Frequently Bought Together
VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Optical bottom plates, 96-well, Nunclon™Δ
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Microplate tilting stand, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
ROTILABO-CENTRIFUGE TUBES 50ML 1 * 500 items
Supplier: Roth Carl
Microplates, 384-well
Supplier: Greiner Bio-One
BOTTLE PET 250ML SQUARE STERILE DOSED THIO 18MG 1 * 297 items
Supplier: LAB3 OWN PRODUCTION
Transfer pipettes, fine tip
Supplier: Alpha Laboratories
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERLIN
Centrifuge tubes with flat caps, MetalFree®
Supplier: Labcon
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
PARTITION INSERTS FOR CRYOBOX 10X10 1 * 10 items
Supplier: Roth Carl
CRYOBOX PP CLEAR 130x130x80 mm 1 * 36 items
Supplier: TENAK
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Supplier: Thermo Fisher Scientific