Order Entry
Netherlands
ContactUsLinkComponent
DIPPMPO ≥98% (by TLC, 1H-NMR, 13C-NMR)
DIPPMPO ≥98% (by TLC, 1H-NMR, 13C-NMR)
  ENZOALX430119M050
 :  
DIPPMPO ≥98% (by TLC, 1H-NMR, 13C-NMR)
  ENZOALX430119M050
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Purity (TLC, ¹H- and ¹³C-NMR):
    ≥98%

 

 

 :
 :263.27 g/mol
 :Ultra-Low Temperature Freezer (–80°C)
 :527704-58-1
 :C₁₁H₂₂NO₄P
 :MFCD08702711