Order Entry
Netherlands
ContactUsLinkComponent
 
5,5-Dimethyl-1-pyrroline-N-oxide ≥98% (by ELSD)
 
undefined
5,5-Dimethyl-1-pyrroline-N-oxide ≥98% (by ELSD)

 

  • Purity (ELSD):
    ≥98%
  • Appearance:
    Pale yellow solid or oil

 

Frequently Bought Together