Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Compound 48/80 trihydrochloride
Compound 48/80 trihydrochloride ≥98%
Compound 48/80 trihydrochloride ≥98%
Compound 48/80 trihydrochloride ≥98%
Catalog # ENZOALX420008M100
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Compound 48/80 trihydrochloride ≥98%
Catalog # ENZOALX420008M100
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
CAS Number:
94724-12-6
Product Details