Order Entry
Netherlands
ContactUsLinkComponent
21-Hydroxyoligomycin A (from Streptomyces sp. AS5351) ≥95% (by HPLC)
21-Hydroxyoligomycin A (from Streptomyces sp. AS5351) ≥95% (by HPLC)
Catalog # ENZOALX380224M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
21-Hydroxyoligomycin A (from Streptomyces sp. AS5351) ≥95% (by HPLC)
Catalog # ENZOALX380224M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :807.1 g/mol
Storage Temperature:Freezer
CAS Number:102042-09-1
Formula:C₄₅H₇₄O₁₂