Order Entry
Netherlands
ContactUsLinkComponent
Baccatine III ≥95% (by HPLC)
Irritant
Health
Baccatine III ≥95% (by HPLC)
Catalog # ENZOALX351005M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
Health
Baccatine III ≥95% (by HPLC)
Catalog # ENZOALX351005M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥95% (HPLC)
  • Source:
    Semisynthetic from Taxus baccata

Specification Test Results

About this item

 :
 :586.634 g/mol
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:27548-93-2
Formula:C₃₁H₃₈O₁₁