Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Oridonin
Oridonin ≥98% (by HPLC)
Oridonin ≥98% (by HPLC)
ENZOALX350390M050
:
ENZO LIFE SCIENCES
:
Oridonin ≥98% (by HPLC)
ENZOALX350390M050
:
ENZO LIFE SCIENCES
:
Pk:
50 mg
Seq:
2370
Purity:
≥98% (HPLC)
Source:
Isolated from Rabdosia rubescens
:
:
364.4 g/mol
:
GHS08
:
Refrigerator
:
28957-04-2
:
C₂₀H₂₈O₆
:
MFCD00221762