Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Evodiamine
Evodiamine ≥98% (by HPLC)
Evodiamine ≥98% (by HPLC)
Evodiamine ≥98% (by HPLC)
Catalog # ENZOALX350330M050
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Evodiamine ≥98% (by HPLC)
Catalog # ENZOALX350330M050
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
303.365 g/mol
CAS Number:
518-17-2
Formula:
C₁₉H₁₇N₃O
MDL Number:
MFCD06407824
Product Details