Order Entry
Netherlands
Orders LinkContactUsLinkComponent
S14-95

S14-95

0,5 mg
Catalog # ENZOALX350299MC05
Supplier:   Enzo Life Sciences
S14-95
Catalog # ENZOALX350299MC05
Supplier:  Enzo Life Sciences
CAS Number:  419532-92-6
S14-95

0,5 mg

CAS Number:  419532-92-6
Price
322,00 €
Log in for availability
1 * 0,5 mg
Product Details & Documents

STAT1alpha inhibitor

Potent inhibitor of cytokine-induced activation of STAT1α leading to the inhibition of inducible expression of proinflammatory enzymes (COX-2, iNOS (NOS II)) and cytokines (TNF-α).

Isolated from Penicillium sp.

  • CAS Number:419532-92-6
  • Formula:C₂₈H₃₄O₅
  • Molecular Weight:450.57 g/mol
Specifications

Specifications

  • Pk:
    0,5 mg

Specifications