Order Entry
Netherlands
ContactUsLinkComponent
Meleagrin (from Penicillium meleagrinum) ≥98% (by HPLC)
Meleagrin (from Penicillium meleagrinum) ≥98% (by HPLC)
Catalog # ENZOALX350297M001
Supplier:  Enzo Life Sciences
CAS Number:  
Meleagrin (from Penicillium meleagrinum) ≥98% (by HPLC)
Catalog # ENZOALX350297M001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :433.5 g/mol
Storage Temperature:Freezer
CAS Number:71751-77-4
Formula:C₂₃H₂₃N₅O₄