Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Xanthohumol
Xanthohumol ≥98% (by HPLC)
Xanthohumol ≥98% (by HPLC)
Xanthohumol ≥98% (by HPLC)
Catalog # ENZOALX350280M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Xanthohumol ≥98% (by HPLC)
Catalog # ENZOALX350280M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
354.40 g/mol
CAS Number:
6754-58-1
Formula:
C₂₁H₂₂O₅
MDL Number:
MFCD00210576
Product Details