

Isoxanthohumol ≥98% (by 1H-NMR)
Catalog # ENZOALX350279M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Purity:≥98% (¹H-NMR)
- Identity:Determined by ¹H-NMR, ¹³C-NMR and LC-MS
- Source:Synthetic
Specification Test Results
About this item
:
:354.40 g/mol
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:70872-29-6
:521-48-2
Formula:C₂₁H₂₂O₅
MDL Number:MFCD00238718