DL-sulphoraphane (synthetic) ≥98% (by UPLC)
Catalog # ENZOALX350232M025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:177.3 g/mol
Storage Temperature:Freezer
CAS Number:4478-93-7
Formula:C₆H₁₁NOS₂
MDL Number:MFCD00198068