D-myo-Inositol-1,3,4,5,6-pentakisphosphate pentapotassium salt ≥98% (by HPLC)
Catalog # ENZOALX307037M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by ¹H-NMR, ³¹P-NMR and MS
Specification Test Results
About this item
:
:770.5 g/mol
Storage Temperature:Freezer
Formula:C₆H₁₂O₂₁P₅·5K