Compound 56 ≥97% (by HPLC)
Catalog # ENZOALX270316C500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:500 µG
- Seq:1418
Specifications
About this item
:
:388.3 g/mol
Storage Temperature:Freezer
CAS Number:171745-13-4
Formula:C₁₈H₁₈BrN₃O₂