6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥96.5% (by titrimetric analysis)
Catalog # ENZOALX270267M100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:250.29 g/mol
Storage Temperature:Refrigerator
CAS Number:53188-07-1
Formula:C₁₄H₁₈O₄
MDL Number:MFCD00006846