Order Entry
Netherlands
Orders LinkContactUsLinkComponent
BML-279

BML-279

5 mg
Catalog # ENZOBMLST3720005
Supplier:   Enzo Life Sciences
BML-279
Catalog # ENZOBMLST3720005
Supplier:  Enzo Life Sciences
CAS Number:  
BML-279
BML-279

5 mg

CAS Number:  
Price
114,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

PLD inhibitor.

Dual phospholipase D1/D2 (PLD1/PLD2) inhibitor in cellular assays (PLD1 IC50=8 nm in Calu-1 cells; PLD2 IC50=42 nm in HEK293-gfpPLD2 cells).

  • Formula:C₂₄H₂₅N₅O₂
  • Molecular Weight:415.5 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.