Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

BML-267

50 mg
Catalog # ENZOBMLPR1210050
Supplier:   Enzo Life Sciences
BML-267
Catalog # ENZOBMLPR1210050
Supplier:  Enzo Life Sciences
CAS Number:  
BML-267
BML-267

50 mg

CAS Number:  
Price
603,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

PTP1B inhibitor

Potent and selective protein tyrosine phosphatase 1B (PTP1B) inhibitor (IC50=11 µM).

  • Formula:C₁₁H₁₂N₂O₅S·C₂HF₃O₂
  • Molecular Weight:398.3 g/mol
Specifications

Specifications

  • Pk:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.