Order Entry
Netherlands
Orders LinkContactUsLinkComponent
BML-282
5 mg
  ENZOBMLEI4000005
BML-282
  ENZOBMLEI4000005
 :  
BML-282
BML-282
5 mg
 :  
165,00 €
1 * 5 mg
UCH inhibitor
Novel inhibitor of ubiquitin C-terminal hydrolase L1 (IC50 = 0.81 µM). It displays 21-fold greater selectivity over UCH-L3 (IC50 = 17 µM).
  •  :C₁₇H₁₁BrCl₂N₂O₃
  •  :442.1 g/mol
  • Pk:
    5 mg